Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696341.3 | XP_005696398.1 | 2 - 3 | 1204 | 69 | |
| XM_013973797.2 | XP_013829251.1 | 2 - 3 | 1207 | 69 | ||
| GenBank mRNA | JO595943.2 | - | 1 - 2 | 2052 | 69 | |
More Information
Set Membership: |
|

MAD2L1BP
Genomic Map