Assay Details
| Gene Symbol: PYROXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698310.3 | XP_005698367.2 | 3 - 4 | 306 | 57 | |
| XM_005698311.3 | XP_005698368.2 | 2 - 3 | 266 | 57 | ||
| GenBank mRNA | JO429118.2 | - | 13 - 14 | 1956 | 57 | |
More Information
Set Membership: |
|

PYROXD2
Genomic Map