Assay Details
| Gene Symbol: LOC102179346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698319.3 | XP_005698376.1 | 4 - 5 | 680 | 103 | |
| GenBank mRNA | JO434318.2 | - | 4 - 5 | 691 | 103 | |
| KA306325.1 | - | 5 - 6 | 2939 | 103 | ||
More Information
Set Membership: |
|

LOC102179346
Genomic Map