Assay Details
| Gene Symbol: LOC102179346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698319.3 | XP_005698376.1 | 7 - 8 | 1097 | 96 | |
| GenBank mRNA | JO434318.2 | - | 7 - 8 | 1108 | 96 | |
| KA306325.1 | - | 2 - 3 | 2522 | 96 | ||
More Information
Set Membership: |
|

LOC102179346
Genomic Map