Assay Details
| Gene Symbol: LOC102179346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698319.3 | XP_005698376.1 | 8 - 9 | 1216 | 67 | |
| GenBank mRNA | KA306325.1 | - | 1 - 2 | 2403 | 67 | |
More Information
Set Membership: |
|

LOC102179346
Genomic Map