Assay Details
| Gene Symbol: CHORDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043296.1 | XP_017898785.1 | 8 - 9 | 798 | 82 | |
| GenBank mRNA | JO581233.1 | - | 3 - 4 | 762 | 82 | |
| JO624029.2 | - | 3 - 4 | 575 | 82 | ||
More Information
Set Membership: |
|

CHORDC1
Genomic Map