Assay Details
| Gene Symbol: PRCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699483.3 | XP_005699540.1 | 4 - 5 | 560 | 76 | |
| XM_005699487.3 | XP_005699544.1 | 4 - 5 | 583 | 76 | ||
| XM_018043431.1 | XP_017898920.1 | 4 - 5 | 733 | 76 | ||
| XM_018043432.1 | XP_017898921.1 | 3 - 4 | 258 | 76 | ||
| GenBank mRNA | JO361773.2 | - | 3 - 4 | 433 | 76 | |
| KA326449.1 | - | 4 - 5 | 716 | 76 | ||
More Information
Set Membership: |
|

PRCP
Genomic Map