Assay Details
| Gene Symbol: LOC108633300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044709.1 | XP_017900198.1 | 4 - 5 | 639 | 74 | |
| GenBank mRNA | JO641536.2 | - | 568 | 74 | ||
More Information
Set Membership: |
|

LOC108633300
Genomic Map