Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044329.1 | XP_017899818.1 | 1145 | 93 | ||
| GenBank mRNA | JT737079.1 | - | 8 - 9 | 1822 | 93 | |
More Information
Set Membership: |
|

GABRA3
Genomic Map