Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060048.1 | XP_017915537.1 | 3 - 4 | 621 | 54 | |
| XM_018060049.1 | XP_017915538.1 | 3 - 4 | 1008 | 54 | ||
| GenBank mRNA | JO342579.2 | - | 3 - 4 | 630 | 54 | |
| JO608524.2 | - | 3 - 4 | 482 | 54 | ||
More Information
Set Membership: |
|

NUAK2
Genomic Map