Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 32 - 33 | 5760 | 119 | |
| GenBank mRNA | JO426300.1 | - | 13 - 14 | 1708 | 119 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map