Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 43 - 44 | 7081 | 80 | |
| GenBank mRNA | JO426300.1 | - | 2 - 3 | 387 | 80 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map