Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 44 - 45 | 7206 | 88 | |
| GenBank mRNA | JO426300.1 | - | 1 - 2 | 262 | 88 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map