Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 23 - 24 | 4086 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102172692| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 23 - 24 | 4086 | 112 | |
Set Membership: |
|