Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 26 - 27 | 4647 | 103 | |
| GenBank mRNA | JO426300.1 | - | 19 - 20 | 2821 | 103 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map