Assay Details
| Gene Symbol: FBXO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048222.1 | XP_017903711.1 | 8 - 9 | 1826 | 66 | |
| XM_018048223.1 | XP_017903712.1 | 8 - 9 | 1041 | 66 | ||
| GenBank mRNA | JO639272.2 | - | 9 - 10 | 1331 | 66 | |
More Information
Set Membership: |
|

FBXO7
Genomic Map