Assay Details
| Gene Symbol: PRKCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967227.2 | XP_013822681.2 | 11 - 12 | 2247 | 54 | |
| XM_018054083.1 | XP_017909572.1 | 11 - 12 | 1938 | 54 | ||
| GenBank mRNA | JO330919.2 | - | 11 - 12 | 1550 | 54 | |
More Information
Set Membership: |
|

PRKCH
Genomic Map