Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057652.1 | XP_017913141.1 | 2 - 3 | 1029 | 62 | |
| XM_018057653.1 | XP_017913142.1 | 2 - 3 | 1023 | 62 | ||
| GenBank mRNA | JT698784.1 | - | 3 - 4 | 692 | 62 | |
More Information
Set Membership: |
|

ANGPT4
Genomic Map