Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970630.2 | XP_013826084.1 | 2 - 3 | 367 | 65 | |
| XM_013970631.2 | XP_013826085.1 | 2 - 3 | 439 | 65 | ||
| XM_013970632.2 | XP_013826086.1 | 2 - 3 | 305 | 65 | ||
More Information
Set Membership: |
|

PRKAB1
Genomic Map