Assay Details
| Gene Symbol: DPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062178.1 | XP_017917667.1 | 4 - 5 | 632 | 70 | |
| XM_018062179.1 | XP_017917668.1 | 5 - 6 | 1100 | 70 | ||
| XM_018062180.1 | XP_017917669.1 | 4 - 5 | 632 | 70 | ||
| XM_018062181.1 | XP_017917670.1 | 2 - 3 | 319 | 70 | ||
| GenBank mRNA | JO608323.2 | - | 4 - 5 | 638 | 70 | |
| JO608324.2 | - | 4 - 5 | 638 | 70 | ||
More Information
Set Membership: |
|

DPEP2
Genomic Map