Assay Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039642.1 | XP_017895131.1 | 3 - 4 | 603 | 62 | |
| GenBank mRNA | JO376750.2 | - | 3 - 4 | 1384 | 62 | |
More Information
Set Membership: |
|

RAB12
Genomic Map