Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040895.1 | XP_017896384.1 | 4 - 5 | 1282 | 69 | |
| XM_018040896.1 | XP_017896385.1 | 2 - 3 | 1202 | 69 | ||
| XM_018040897.1 | XP_017896386.1 | 3 - 4 | 1165 | 69 | ||
| XM_018040898.1 | XP_017896387.1 | 4 - 5 | 1282 | 69 | ||
| GenBank mRNA | JO610455.2 | - | 3 - 4 | 1151 | 69 | |
More Information
Set Membership: |
|

PKMYT1
Genomic Map