Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674814.3 | XP_005674871.2 | 9 - 10 | 1283 | 100 | |
| XM_018057058.1 | XP_017912547.1 | 9 - 10 | 1356 | 100 | ||
| XM_018057168.1 | XP_017912657.1 | 9 - 10 | 1353 | 100 | ||
| GenBank mRNA | JO354595.2 | - | 1 - 2 | 100 | ||
| JO378529.2 | - | 1 - 2 | 100 | |||
| JO399491.2 | - | 8 - 9 | 1212 | 100 | ||
| JO433425.2 | - | 1 - 2 | 100 | |||
| JO594009.2 | - | 4 - 5 | 423 | 100 | ||
| JO618219.2 | - | 1 - 2 | 100 | |||
| JT716509.1 | - | 3 - 4 | 350 | 100 | ||
| KA331185.1 | - | 1 - 2 | 100 | |||
More Information
Set Membership: |
|

RIOX2
Genomic Map