Assay Details
| Gene Symbol: LOC102186637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675805.3 | XP_005675862.2 | 60 - 61 | 9618 | 72 | |
| XM_018066077.1 | XP_017921566.1 | 60 - 61 | 9615 | 72 | ||
| XM_018066078.1 | XP_017921567.1 | 60 - 61 | 9603 | 72 | ||
More Information
Set Membership: |
|

LOC102186637
Genomic Map