Assay Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676009.1 | XP_005676066.1 | 2 - 3 | 404 | 94 | |
| XM_005676010.1 | XP_005676067.1 | 2 - 3 | 404 | 94 | ||
More Information
Set Membership: |
|

G6PC2
Genomic Map