Assay Details
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676268.3 | XP_005676325.1 | 5 - 6 | 717 | 70 | |
GenBank mRNA | JO581875.1 | - | 5 - 6 | 812 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
