Assay Details
| Gene Symbol: PLA2G2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676914.3 | XP_005676971.2 | 4 - 5 | 412 | 91 | |
| GenBank mRNA | JO591235.2 | - | 1 - 2 | 538 | 91 | |
More Information
Set Membership: |
|

PLA2G2A
Genomic Map