Assay Details
| Gene Symbol: AREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681761.3 | XP_005681818.3 | 4 - 5 | 844 | 73 | |
| XM_013964723.2 | XP_013820177.2 | 4 - 5 | 1065 | 73 | ||
| XM_018049584.1 | XP_017905073.1 | 5 - 6 | 817 | 73 | ||
| XM_018049585.1 | XP_017905074.1 | 5 - 6 | 817 | 73 | ||
More Information
Set Membership: |
|

AREG
Genomic Map