Assay Details
| Gene Symbol: LOC102178535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050351.1 | XP_017905840.1 | 4 - 5 | 704 | 82 | |
| XM_018050352.1 | XP_017905841.1 | 4 - 5 | 522 | 82 | ||
| XM_018050353.1 | XP_017905842.1 | 4 - 5 | 522 | 82 | ||
| XM_018050355.1 | XP_017905844.1 | 4 - 5 | 705 | 82 | ||
More Information
Set Membership: |
|

LOC102178535
Genomic Map