Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052512.1 | XP_017908001.1 | 3 - 4 | 541 | 58 | |
| GenBank mRNA | JO545679.2 | - | 1 - 2 | 490 | 58 | |
More Information
Set Membership: |
|

GADD45G
Genomic Map