Assay Details
| Gene Symbol: EMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685453.3 | XP_005685510.1 | 1 - 2 | 328 | 80 | |
| GenBank mRNA | JO580478.1 | - | 4 - 5 | 864 | 80 | |
More Information
Set Membership: |
|

EMC7
Genomic Map