Assay Details
| Gene Symbol: TRAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059227.1 | XP_017914716.1 | 7 - 8 | 1074 | 70 | |
| XM_018059228.1 | XP_017914717.1 | 6 - 7 | 979 | 70 | ||
| GenBank mRNA | JO589366.2 | - | 6 - 7 | 956 | 70 | |
| JQ308791.1 | - | 5 - 6 | 589 | 70 | ||
| KA339914.1 | - | 1 - 2 | 970 | 70 | ||
More Information
Set Membership: |
|

TRAF6
Genomic Map