Assay Details
| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690390.3 | XP_005690447.1 | 2 - 3 | 224 | 69 | |
| XM_005690392.3 | XP_005690449.1 | 2 - 3 | 221 | 69 | ||
| XM_013970022.2 | XP_013825476.1 | 2 - 3 | 224 | 69 | ||
| XM_018060017.1 | XP_017915506.1 | 2 - 3 | 224 | 69 | ||
| GenBank mRNA | JO593084.2 | - | 9 - 10 | 1572 | 69 | |
More Information
Set Membership: |
|

MDM4
Genomic Map