Assay Details
| Gene Symbol: GORAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690632.3 | XP_005690689.2 | 4 - 5 | 766 | 55 | |
| GenBank mRNA | JT709017.1 | - | 5 - 6 | 718 | 55 | |
More Information
Set Membership: |
|

GORAB
Genomic Map