Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691822.3 | XP_005691879.2 | 9 - 10 | 1065 | 72 | |
| XM_005691823.3 | XP_005691880.2 | 9 - 10 | 1173 | 72 | ||
| XM_018061756.1 | XP_017917245.1 | 9 - 10 | 902 | 72 | ||
| GenBank mRNA | JT691850.1 | - | 8 - 9 | 836 | 72 | |
More Information
Set Membership: |
|

CENPN
Genomic Map