Assay Details
| Gene Symbol: CDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691833.2 | XP_005691890.2 | 2 - 3 | 284 | 65 | |
| XM_013970937.2 | XP_013826391.2 | 2 - 3 | 461 | 65 | ||
| GenBank mRNA | KA310467.1 | - | 10 - 11 | 1553 | 65 | |
More Information
Set Membership: |
|

CDH13
Genomic Map