Assay Details
| Gene Symbol: VPS4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062209.1 | XP_017917698.1 | 10 - 11 | 1394 | 61 | |
| XM_018062210.1 | XP_017917699.1 | 10 - 11 | 1391 | 61 | ||
| GenBank mRNA | JT763376.1 | - | 1 - 2 | 845 | 61 | |
More Information
Set Membership: |
|

VPS4A
Genomic Map