Assay Details
| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065610.1 | XP_017921099.1 | 4 - 5 | 379 | 66 | |
| XM_018065611.1 | XP_017921100.1 | 4 - 5 | 774 | 66 | ||
| GenBank mRNA | KA305375.1 | - | 3 - 4 | 287 | 66 | |
More Information
Set Membership: |
|

TENT4A
Genomic Map