Assay Details
| Gene Symbol: HIGD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066905.1 | XP_017922394.1 | 3 - 4 | 372 | 103 | |
| XM_018066906.1 | XP_017922395.1 | 3 - 4 | 330 | 103 | ||
| GenBank mRNA | JO636577.2 | - | 3 - 4 | 357 | 103 | |
More Information
Set Membership: |
|

HIGD1A
Genomic Map