Assay Details
| Gene Symbol: ASAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042035.1 | XP_017897524.1 | 8 - 9 | 745 | 64 | |
| GenBank mRNA | JO330007.2 | - | 8 - 9 | 705 | 64 | |
More Information
Set Membership: |
|

ASAH1
Genomic Map