Assay Details
| Gene Symbol: HDHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048703.1 | XP_017904192.1 | 3 - 4 | 448 | 61 | |
| XM_018048704.1 | XP_017904193.1 | 3 - 4 | 557 | 61 | ||
| GenBank mRNA | JO608219.2 | - | 3 - 4 | 431 | 61 | |
| KA321979.1 | - | 2 - 3 | 283 | 61 | ||
More Information
Set Membership: |
|

HDHD5
Genomic Map