Assay Details
| Gene Symbol: LRRC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050256.1 | XP_017905745.1 | 7 - 8 | 1051 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC34| Gene Symbol: LRRC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050256.1 | XP_017905745.1 | 7 - 8 | 1051 | 69 | |
Set Membership: |
|