Assay Details
| Gene Symbol: HCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050508.1 | XP_017905997.1 | 7 - 8 | 2056 | 65 | |
| GenBank mRNA | JO632303.2 | - | 1 - 2 | 1338 | 65 | |
More Information
Set Membership: |
|

HCN2
Genomic Map