Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966746.2 | XP_013822200.1 | 3 - 4 | 1163 | 62 | |
| XM_018053653.1 | XP_017909142.1 | 3 - 4 | 1163 | 62 | ||
| GenBank mRNA | JO609082.2 | - | 3 - 4 | 576 | 62 | |
| KA325857.1 | - | 3 - 4 | 1106 | 62 | ||
More Information
Set Membership: |
|

PARP16
Genomic Map