Assay Details
| Gene Symbol: BCL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055276.1 | XP_017910765.1 | 1 - 2 | 575 | 68 | |
| XM_018055277.1 | XP_017910766.1 | 1 - 2 | 575 | 68 | ||
| XM_018055279.1 | XP_017910768.1 | 1 - 2 | 561 | 68 | ||
More Information
Set Membership: |
|

BCL11A
Genomic Map