Assay Details
| Gene Symbol: PXMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061457.1 | XP_017916946.1 | 2 - 3 | 432 | 59 | |
| GenBank mRNA | JO635598.2 | - | 3 - 4 | 347 | 59 | |
More Information
Set Membership: |
|

PXMP2
Genomic Map