Assay Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040033.1 | XP_017895522.1 | 8 - 9 | 1123 | 59 | |
| XM_018040034.1 | XP_017895523.1 | 8 - 9 | 1333 | 59 | ||
More Information
Set Membership: |
|

CRAMP1
Genomic Map