Assay Details
| Gene Symbol: ACVR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679946.3 | XP_005680003.1 | 8 - 9 | 1488 | 66 | |
| XM_013963787.2 | XP_013819241.1 | 8 - 9 | 1515 | 66 | ||
| XM_018047738.1 | XP_017903227.1 | 8 - 9 | 1443 | 66 | ||
More Information
Set Membership: |
|

ACVR1B
Genomic Map