Assay Details
| Gene Symbol: MRPL55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682073.3 | XP_005682130.1 | 3 - 4 | 641 | 96 | |
| XM_013964947.2 | XP_013820401.1 | 4 - 5 | 716 | 96 | ||
| XM_013964948.2 | XP_013820402.1 | 4 - 5 | 713 | 96 | ||
| XM_018051588.1 | XP_017907077.1 | 4 - 5 | 535 | 96 | ||
| XM_018051589.1 | XP_017907078.1 | 4 - 5 | 532 | 96 | ||
| XM_018051590.1 | XP_017907079.1 | 3 - 4 | 638 | 96 | ||
| XM_018051591.1 | XP_017907080.1 | 3 - 4 | 466 | 96 | ||
| XM_018051592.1 | XP_017907081.1 | 3 - 4 | 463 | 96 | ||
| GenBank mRNA | JO638584.2 | - | 1 - 2 | 337 | 96 | |
| KA329441.1 | - | 3 - 4 | 464 | 96 | ||
More Information
Set Membership: |
|

MRPL55
Genomic Map