Assay Details
| Gene Symbol: ITGB1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055687.1 | XP_017911176.1 | 4 - 5 | 439 | 63 | |
| XM_018055688.1 | XP_017911177.1 | 4 - 5 | 407 | 63 | ||
| GenBank mRNA | KA333704.1 | - | 4 - 5 | 380 | 63 | |
More Information
Set Membership: |
|

ITGB1BP1
Genomic Map